Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Component C3aR, Mouse anti-Human, Clone: 3G7, Novus Biologicals™

Mouse Monoclonal Antibody

$135.50 - $419.00

Specifications

Antigen Complement Component C3aR
Clone 3G7
Applications Flow Cytometry, ELISA
Host Species Mouse
Target Species Human
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB015156
View Documents
Novus Biologicals
NBP2810620.025MG
0.025 mg
Each for $135.50
Add to cart
 
NB015157
View Documents
Novus Biologicals
NBP2810620.2MG
0.2 mg
Each for $419.00
Add to cart
 
Description

Description

Complement Component C3aR Monoclonal antibody specifically detects Complement Component C3aR in Human Samples. It is validated for Flow Cytometry, ELISA.
Specifications

Specifications

Complement Component C3aR
Flow Cytometry, ELISA
Human
Primary
3G7
Mouse
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit