Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Complement Component C5/C5a Antibody Pair [HRP]

Mouse Monoclonal Antibody Pair

$629.00

Specifications

Target Complement Component C5/C5a
Conjugate HRP
Target Species Human
Gene ID (Entrez) 727
Gene Symbol C5
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB010391
View Documents
Novus Biologicals™
NBP27967615PLATES
15 Plates
Each for $629.00
Add to cart
 
NB010392
View Documents
Novus Biologicals™
NBP2796765PLATES
5 Plates This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Complement Component C5/C5a.
Specifications

Specifications

Complement Component C5/C5a
Human
C5
HRP
727
Mouse
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit