Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Component C5a Rabbit anti-Human, Clone: 442, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | Complement Component C5a |
---|---|
Clone | 442 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
Complement Component C5a Monoclonal specifically detects Complement Component C5a in Human samples. It is validated for ELISA.Specifications
Complement Component C5a | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
727 | |
IgG | |
Protein A purified |
442 | |
ELISA | |
Unconjugated | |
Rabbit | |
Human | |
C5, C5A, HE | |
Recombinant Human Complement Component C5a protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title