Learn More
Complement Component C9 Mouse anti-Human, Alexa Fluor™ 594, Clone: 22, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP221612AF594
Catalog No. NP221612A94
Add to cart
Description
Description
Complement Component C9 Monoclonal antibody specifically detects Complement Component C9 in Human samples. It is validated for Western Blot,ELISA,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunofluorescence.
Specifications
Specifications
Complement Component C9
22
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunofluorescence
complement component 9, complement component C9
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
C9
Complement factor C9 adsorbed onto aluminum hydroxide gel.
0.1 mL
Primary
735