Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Factor B Mouse anti-Human, Alexa Fluor™ 350, Clone: 9B8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223508AF350

Catalog No. NP22308A30


Add to cart

Description

Description

Complement Factor B Monoclonal antibody specifically detects Complement Factor B in Human samples. It is validated for Western Blot,ELISA.
Specifications

Specifications

Complement Factor B
9B8
Western Blot, ELISA
BF AHUS4, B-factor, properdin, BFD, C3 proaccelerator, C3 proactivator, C3/C5 convertase, complement factor B, EC 3.4.21, EC 3.4.21.47, FB, FBI12, GBGCFAB, Glycine-rich beta glycoprotein, glycine-rich beta-glycoprotein, H2-Bf, PBF2, Properdin factor B
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Clone 9B8 is specific for the Bb fragment of human factor B. Antibody binding abolishes the formation of C3bBb complexes.
ELISA, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
CFB
Complement factor B purified from human plasma and adsorbed to aluminum hydroxide gel
0.1 mL
Primary
629
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit