Learn More
Complement Factor D/Adipsin Mouse anti-Human, Alexa Fluor™ 350, Clone: MM0196-4M47, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212203AF350
Catalog No. NP212203A30
Add to cart
Description
Description
Complement Factor D/Adipsin Monoclonal antibody specifically detects Complement Factor D/Adipsin in Human samples. It is validated for Western Blot.
Specifications
Specifications
Complement Factor D/Adipsin
MM0196-4M47
Western Blot
Adipsin, ADNcomplement factor D, C3 convertase activator, complement factor D (adipsin), complement factor D preproprotein, D component of complement (adipsin), DF, EC 3.4.21, EC 3.4.21.46, PFD, Properdin factor DADIPSIN
Mouse
IgG2
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
CFD
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with human recombinant protein of Adipsin.
0.1 mL
Primary
1675