Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Factor D/Adipsin Mouse anti-Human, Alexa Fluor™ 532, Clone: D10/4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223532AF532

Catalog No. NP22332A32


Add to cart

Description

Description

Complement Factor D/Adipsin Monoclonal antibody specifically detects Complement Factor D/Adipsin in Human samples. It is validated for Western Blot,ELISA.
Specifications

Specifications

Complement Factor D/Adipsin
D10/4
Western Blot, ELISA
Adipsin, ADNcomplement factor D, C3 convertase activator, complement factor D (adipsin), complement factor D preproprotein, D component of complement (adipsin), DF, EC 3.4.21, EC 3.4.21.46, PFD, Properdin factor DADIPSIN
Mouse
IgG2a Kappa
Store at 4°C in the dark.
Monoclonal
Clone D10/4 is specific for complement factor D in human serum and plasma
ELISA, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CFD
Human Factor D
0.1 mL
Primary
1675
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit