Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Factor H Mouse anti-Human, Alexa Fluor™ 532, Clone: C18/3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP223542AF532

Catalog No. NP22342A32


Add to cart

Description

Description

Complement Factor H Monoclonal antibody specifically detects Complement Factor H in Human samples. It is validated for Western Blot,ELISA,Immunohistochemistry.
Specifications

Specifications

Complement Factor H
C18/3
Western Blot, ELISA, Immunohistochemistry
adrenomedullin binding protein, age-related maculopathy susceptibility 1, AHUS1, AMBP1, ARMD4, beta-1H, beta-1-H-globulin, CFHL3, complement factor H, factor H, factor H-like 1, FH, FHL1, H factor 1, H factor 1 (complement), H factor 2 (complement), HF, HF1ARMS1, HF2, HUSMGC88246
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Clone C18/3 is specific for complement factor H in human serum and plasma
ELISA, Immunohistochemistry, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CFH
Human Factor H
0.1 mL
Primary
3075
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit