Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Factor H Mouse anti-Human, Clone: C18/3, Novus Biologicals™
Mouse Monoclonal Antibody
$177.31 - $436.00
Specifications
Antigen | Complement Factor H |
---|---|
Clone | C18/3 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB23542100U
|
Novus Biologicals
NBP223542100UG |
100 μg |
Each for $436.00
|
|
NB019323
|
Novus Biologicals
NBP2235420.025MG |
0.025 mg |
Each for $177.31
|
|
Description
Complement Factor H Monoclonal specifically detects Complement Factor H in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
Complement Factor H | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Human | |
adrenomedullin binding protein, age-related maculopathy susceptibility 1, AHUS1, AMBP1, ARMD4, beta-1H, beta-1-H-globulin, CFHL3, complement factor H, factor H, factor H-like 1, FH, FHL1, H factor 1, H factor 1 (complement), H factor 2 (complement), HF, HF1ARMS1, HF2, HUSMGC88246 | |
CFH | |
IgG1 κ | |
Protein A or G purified | |
Clone C18/3 is specific for complement factor H in human serum and plasma |
C18/3 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide | |
3075 | |
Human Factor H | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title