Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Factor H Rabbit anti-Human, Clone: 305, Novus Biologicals™
Rabbit Monoclonal Antibody
$340.00 - $521.00
Specifications
Antigen | Complement Factor H |
---|---|
Clone | 305 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
Complement Factor H Monoclonal specifically detects Complement Factor H in Human samples. It is validated for ELISA.Specifications
Complement Factor H | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
3075 | |
IgG | |
Protein A purified |
305 | |
ELISA | |
Unconjugated | |
Rabbit | |
Human | |
adrenomedullin binding protein, age-related maculopathy susceptibility 1, AHUS1, AMBP1, ARMD4, beta-1H, beta-1-H-globulin, CFHL3, complement factor H, factor H, factor H-like 1, FH, FHL1, H factor 1, H factor 1 (complement), H factor 2 (complement), HF, HF1ARMS1, HF2, HUSMGC88246 | |
Recombinant Human Complement Factor H protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title