Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement factor H Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | Complement factor H |
---|---|
Concentration | 0.35 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8218
![]() ![]() |
Proteintech Group Inc
127481AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Factor H is a regulatory factor of the alternative pathway of complement activation. Factor H is a glycoprotein consisting of a single polypeptide chain with a molecular mass of 150 kDa.Specifications
Complement factor H | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P08603 | |
3075 | |
Complement factor H Fusion Protein Ag3482 | |
Primary | |
-20°C | |
CFH |
0.35 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ARMD4, ARMS1, CFH, CFHL3, Complement factor H, Factor H, FH, FHL1, H factor 1, HF, HF1, HF2, HUS | |
CFH | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title