Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Factor I Mouse anti-Human, Alexa Fluor™ 750, Clone: 3D6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP223564AF750

Catalog No. NP22364A70

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Complement Factor I Monoclonal antibody specifically detects Complement Factor I in Human samples. It is validated for Western Blot,ELISA.
Specifications

Specifications

Complement Factor I
3D6
Western Blot, ELISA
AHUS3, C3B/C4B inactivator, C3BINA, C3b-inactivator, complement component I, complement control protein factor I, complement factor I, complement factor I heavy chain, EC 3.4.21, EC 3.4.21.45, FIC3b-INA, I factor (complement), IF, KAF, Konglutinogen-activating factor, light chain of factor I
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Clone 3D6 is specific for the alpha-chain of human factor I
ELISA, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
CFI
Factor I isolated from human plasma adsorbed onto aluminum hydroxide gel
0.1 mL
Primary
3426
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit