Learn More
Complement receptor 1 Mouse anti-Human, Unconjugated, Clone: 7G9, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma MABF2239100UL
 View more versions of this product
Catalog No. MABF22391MI
Add to cart
Description
Description
Complement receptor 1 Monoclonal antibody specifically detects Complement receptor 1 in Human,Rhesus Monkey samples. It is validated for Dot Blot, Flow Cytometry, Radioimmunoassay.
Specifications
Specifications
Complement receptor 1
7G9
NP_000564.2
C3b/C4b receptor;CD35
Mouse
IgG2aκ
Protein G Purified
Inflammation and Immunology
Primary
Clone 7G9 is a mouse monoclonal antibody that detects human complement receptor 1 (CR1). it binds to long homologous repeats (LHR) A,B,C of CR1.
Dot Blot, Flow Cytometry, Radioimmune Assays
Unconjugated
Purified
CR1, C3BR
Soluble human complement receptor 1.
223.66 kDa calculated.
100 μL
Stable for 1 year at 2 to 8°C from date of receipt.
Monoclonal
Human, Rhesus Monkey