Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement receptor 1 Mouse anti-Human, Unconjugated, Clone: 7G9, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABF223925UL
Description
Complement receptor 1 Monoclonal antibody specifically detects Complement receptor 1 in Human,Rhesus Monkey samples. It is validated for Dot Blot, Flow Cytometry, Radioimmunoassay.Specifications
Complement receptor 1 | |
Monoclonal | |
Unconjugated | |
C3b/C4b receptor;CD35 | |
Mouse | |
223.66 kDa calculated. | |
25 μL | |
Primary | |
Human, Rhesus Monkey | |
Purified |
Dot Blot, Flow Cytometry, Radioimmune Assays (RIA) | |
7G9 | |
NP_000564.2 | |
CR1, C3BR | |
Soluble human complement receptor 1. | |
Protein G Purified | |
Inflammation and Immunology | |
Clone 7G9 is a mouse monoclonal antibody that detects human complement receptor 1 (CR1). it binds to long homologous repeats (LHR) A,B,C of CR1. | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction