Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement receptor 1 Mouse anti-Human, Unconjugated, Clone: 7G9, MilliporeSigma™

Mouse Monoclonal Antibody

$153.50 - $386.00

Specifications

Antigen Complement receptor 1
Applications Dot Blot, Flow Cytometry, Radioimmune Assays
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF22391MI
View Documents
MilliporeSigma
MABF2239100UL
100 μL
Each for $386.00
Add to cart
 
MABF22392MI
View Documents
MilliporeSigma
MABF223925UL
25 μL
Each for $153.50
Add to cart
 
Description

Description

Complement receptor 1 Monoclonal antibody specifically detects Complement receptor 1 in Human,Rhesus Monkey samples. It is validated for Dot Blot, Flow Cytometry, Radioimmunoassay.
Specifications

Specifications

Complement receptor 1
Unconjugated
Mouse
C3b/C4b receptor;CD35
Soluble human complement receptor 1.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Dot Blot, Flow Cytometry, Radioimmune Assays
Purified
Human, Rhesus Monkey
CR1, C3BR
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit