Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ComplementComponentC2 Mouse anti-Human, Janelia Fluor 549, Clone: 5C3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223549JF549

Catalog No. NBP23549549


Add to cart

Description

Description

ComplementComponentC2 Monoclonal antibody specifically detects ComplementComponentC2 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

ComplementComponentC2
5C3
Western Blot, ELISA, Immunohistochemistry
Purified
C2
Complement component C2 isolated from human plasma adsorbed onto aluminum hydroxide gel
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
Clone 5C3 is specific for C2 from human plasma/serum. Cross-reactivity to C2 from other species has not been tested.
ELISA, Immunohistochemistry, Western Blot
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
C3/C5 convertase, CO2, complement C2, complement component 2, complement component C2, DKFZp779M0311, EC 3.4.21, EC 3.4.21.43
Mouse
IgG2a Kappa
0.1mL
Primary
717
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit