Learn More
GSC Go Science Crazy Complete Cartesian Diver Kit
Cartesian diver demonstrates Archimedes' principle of buoyancy.
$14.75
Specifications
For Use With (Application) | Can be used as replacement to Cartesian Diver Unit or used in other cylinders or soda pop bottles experiments to demonstrates Archimedes' principle of buoyancy |
---|
Includes:
- 38mm Cartesian diver
- Hydrometer cylinder, 24 x 4cm
- Rubber diaphragm
- Instruction sheet with demonstration suggestions
Catalog Number | Mfr. No. | Includes | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Product Type | Price | Quantity & Availability | ||||
S41122
![]() |
GSC Go Science Crazy
3051 |
38mm H Cartesian diver, 24 x 4cm hydrometer cylinder, Rubber diaphragm, instruction sheet with demonstration suggestions | Complete Cartesian diver apparatus |
Each for $14.75
|
|
Description
- Show the transmission of pressure by a liquid, compressibility of gases, and Archimedes' Principle
- Hollow glass diver dips and returns to the surface of the cylinder using a small hole that admits air and water
- Extra divers available separately
Specifications
Can be used as replacement to Cartesian Diver Unit or used in other cylinders or soda pop bottles experiments to demonstrates Archimedes' principle of buoyancy |