Learn More
Complete Optical Bench System Products for Science Education
Quality, long-lasting instruments for use with a laser.
Includes:
  • Two 23.6 in. (0.6m) benches
  • Two leveling base sets
  • Adjustable laser holder
  • Heavy-duty pin carrier with pin
  • Two ring mounts
  • Two adjustable mirror holders
  • Four standard pin carriers with pins
Requires:
  • Separate laser
Products 1
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S42815ND Products for Science Education
View Documents
Industrial Fiber Optics IncSupplier Diversity Partner
45212
Pin carrier This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Education-grade bench system designed with component interchangeability
  • Standard 1/4 in.-20 threads on mounting pins
  • Nearly universal exchangeability with other systems