Learn More
Fisherbrand™ Complete Plumbing Assemblies for Lab Workstations
Fisherbrand™ Complete Plumbing Assemblies are furnished complete for Lab Workstations. Designed specifically for use in the laboratory environment.
Specifications
Material PVC, Chrome
Includes Male quick connect, system hose and service turret
View More Specs
Includes: Male Quick Connect, System Hose and Service Turret
Products 2
Catalog Number dimColor Type Price Quantity & Availability  
Catalog Number dimColor Type Price Quantity & Availability  
06-000-357
View Documents Promotion Details
Vacuum Service, Left Side This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
06-000-360
View Documents Promotion Details
Vacuum Service, Right Side This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Service turret is color coded base on service
  • Service turret is a 135° degreet turret with one polished chrome valve with a brass floating cone and integral brass seat.
  • Service turrets provide metering control of laboratory gases at pressures up to 60 PSI.
  • System service hoses carry services from the overhead service panel through the upright of the table and to the shank of the fixture. Hoses extend 48 in. above the workstation
  • System service hoses are color coded reinforced PVC
  • Male Quick connect: double end shut off plugs have an internal valve that shuts off flow when uncoupled
Specifications
Specifications
PVC, Chrome
Male quick connect, system hose and service turret