Learn More
DWK Life Sciences Wheaton™ CompletePAK Sterile Kit with 10mL Serum Vials, 20mm Stopper, and Flip Seal
Wheaton CompletePAK product line is a range of crimp top vials, rubber stoppers and seals that are ready-to-use and are off-the-shelf, customizable for any application
$1189.16 - $1324.29
Specifications
Closure Size 20mm
Sterility Sterile
No. per Case 158
Product Line Wheaton CompletePAK
Certifications/Compliance USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
View More Specs
Products 6
Catalog Number Mfr. No. Closure Type Cap Style Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Closure Type Cap Style Volume (Metric) Price Quantity & Availability  
06-451-232 View Documents DWK Life Sciences
W990012CP
Ultra Pure Serum Stopper, Blue Flip Seal 10 mL Case of 158 for $1,189.16
Add to cart
06-451-233 View Documents DWK Life Sciences
W990013CP
Omni Flex 3G Straight Plug Stopper, Blue Flip Seal 10 mL Case of 158 for $1,291.86
Add to cart
06-451-234 View Documents DWK Life Sciences
W990014CP
Igloo Lyo Stopper, Blue Flip Seal 10 mL Case of 158 for $1,324.29
Add to cart
06-451-235 View Documents DWK Life Sciences
W990015CP
Ultra Pure Serum Stopper, Red Flip Seal 10 mL Case of 158 for $1,189.16
Add to cart
06-451-236 View Documents DWK Life Sciences
W990016CP
Omni Flex 3G Straight Plug Stopper, Red Flip Seal 10 mL Case of 158 for $1,291.86
Add to cart
06-451-237 View Documents DWK Life Sciences
W990017CP
Igloo Lyo Stopper, Red Flip Seal 10 mL Case of 158 for $1,324.29
Add to cart
Description
Description
  • Type I glass serum vials
  • Sterile 10mL vials
  • Range of sterile stoppers and seals
  • Each component is particulate cleaned, dehydrogenated, and sterilized certified to USP <788>, <85> & <71>
  • All components are Container Closure Integrity (CCI) certified
  • Each kit has 220 stoppers and seals
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
Specifications
Specifications
20mm
158
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
Sterile
Wheaton CompletePAK