Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ CompletePAK Sterile Kit with 2mL Serum Vials, 13mm Stopper, and Flip Seal

Wheaton CompletePAK product line is a range of crimp top vials, rubber stoppers and seals that are ready-to-use and are off-the-shelf, customizable for any application

Manufacturer:  DWK Life Sciences W990005CP

 View more versions of this product

Catalog No. 06-451-225


Add to cart

Description

Description

Wheaton CompletePAK product line is a range of crimp top vials, rubber stoppers and seals that are ready-to-use and are off-the-shelf, customizable for any application

  • Type I glass serum vials
  • Sterile 2mL vials
  • Range of sterile stoppers and seals
  • Each component is particulate cleaned, dehydrogenated, and sterilized certified to USP <788>, <85> & <71>
  • All components are Container Closure Integrity (CCI) certified
  • Each kit has 230 stoppers and seals

Certifications

USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified

Specifications

Specifications

Bromobutyl, Glass
2 mL
Wheaton™ CompletePAK
USP 788, 85, and 71, Container Closure Integrity (CCI) certified
Sterile
210
Igloo Lyo Stopper, Red Flip Seal
13 mm
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit