Learn More
DWK Life Sciences Wheaton™ CompletePAK Sterile Kit with 5mL Serum Vials, 20mm Stopper, and Flip Seal Available on GSA/VA Contract for Federal Government customers only.
Wheaton CompletePAK product line is a range of crimp top vials, rubber stoppers and seals that are ready-to-use and are off-the-shelf, customizable for any application
$1135.10 - $1270.24
Specifications
Closure Size 20mm
Sterility Sterile
Material Butyl, Glass
No. per Case 200
Certifications/Compliance USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
View More Specs
Products 6
Catalog Number Mfr. No. Closure Type Cap Style Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Closure Type Cap Style Volume (Metric) Price Quantity & Availability  
06-451-226 Available on GSA/VA Contract for Federal Government customers only. View Documents DWK Life Sciences
W990006CP
Ultra Pure Serum Stopper, Blue Flip Seal 5 mL Case of 200 for $1,135.10
Add to cart
06-451-227 View Documents DWK Life Sciences
W990007CP
Omni Flex 3G Straight Plug Stopper, Blue Flip Seal 5 mL Case of 200 for $1,237.80
Add to cart
06-451-228 View Documents DWK Life Sciences
W990008CP
Igloo Lyo Stopper, Blue Flip Seal 5 mL Case of 200 for $1,270.24
Add to cart
06-451-229 View Documents DWK Life Sciences
W990009CP
Ultra Pure Serum Stopper, Red Flip Seal 5 mL Case of 200 for $1,135.10
Add to cart
06-451-230 View Documents DWK Life Sciences
W990010CP
Omni Flex 3G Straight Plug Stopper, Red Flip Seal 5 mL Case of 200 for $1,237.80
Add to cart
06-451-231 View Documents DWK Life Sciences
W990011CP
Igloo Lyo Stopper, Red Flip Seal 5 mL Case of 200 for $1,270.24
Add to cart
Description
Description
  • Type I glass serum vials
  • Sterile 5mL vials
  • Range of sterile stoppers and seals
  • Each component is particulate cleaned, dehydrogenated, and sterilized certified to USP <788>, <85> & <71>
  • All components are Container Closure Integrity (CCI) certified
  • Each kit has 220 stoppers and seals
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
20mm
Butyl, Glass
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
Sterile
200