Learn More
Complexin-2 Rabbit anti-Human, Alexa Fluor 405, Clone: 21, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP290019AF405
Catalog No. NB036904
Add to cart
Description
Description
Complexin-2 Monoclonal specifically detects Complexin-2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Complexin-2
21
Flow Cytometry, Immunocytochemistry/Immunofluorescence
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Immunofluorescence
Alexa Fluor 405
50mM Sodium Borate
complexin 2, Complexin II, complexin-2, CPX II, CPX2, CPX-2, DKFZp547D155, Hfb1, MGC138492, synaphin 1,921-L, synaphin-1
This antibody was obtained from a rabbit immunized with purified, recombinant Human Complexin-2 (Q6PUV4-1; Asp2-Lys134).
Protein A purified
RUO
Primary
10814