Learn More
Enzo Life SciencesSupplier Diversity Partner Complement component C3a/C3a (desArg) monoclonal antibody (K13/16) (200 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner BPDGAU0131602
Clone: K13/16. Host: Mouse. Isotype: IgG1κ. Immunogen: Human complement component C3a. UniProt: P01024. Species Reactivity: Human. Specificity: Recognizes C3, C3a and C3a (desArg). Crossreactivity: Does not cross-react with C4a or C5a. Applications: ELISA, IF, IHC, WB. Purity: Protein A/G-affinity purified. Formulation: Liquid. In 0.01M phosphate buffer, pH 7.4, containing 0.5M sodium chloride and 15mM sodium azide.
Catalog No. 50-659-163