Learn More
MEDPLUS SERVICES USA COMPLY INDICAT STRPS 1000/CS
Supplier:  MEDPLUS SERVICES USA MMM 1248
Encompass_Preferred
Internal pack monitor for use with Vaporized Hydrogen Peroxide/Gas Plasma sterilization processes Distinct blue to pink color change Can be validated to other Vaporized Hydrogen Peroxide cycles Blue to pink color change Indicator does not fade when exposed to light
Catalog No. NC0375318
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.