Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Compound 401, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Reversible and selective inhibitor of DNA-dependent protein kinase (DNA-PK) and mammalian target of rapamycin (mTOR) (IC50 values are 0.28 and 5.3 μM respectively). Displays little affinity for other commonly studied kinases including PI 3-K, ATM and ATR (IC50 values are all > 100 μM). Induces apoptosis in vitro.
Specifications
Quantity | 10 mg |
Formula Weight | 281.31 |
Percent Purity | >99% |
Chemical Name or Material | Compound 401 |
Safety and Handling
Recommended Storage : Store at Room Temperature
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title