Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Compound Benzoin Tincture, USP, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum

BE150

Spectrum™ Compound Benzoin Tincture, USP is a pungent solution used as an oral mucosal protectan. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Quantity 500 mL
Packaging Amber Glass Bottle
CAS Max % 100%
UN Number UN1170