Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Compound W, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
173550-33-9
C19H12N2O8
396.311
JOSXKPZXMVHRKU-UHFFFAOYSA-N
3,5-bis 4-nitrophenoxy benzoic acid,gamma-secretase modulator, cw,cw,acmc-1cuip,josxkpzxmvhrku-uhfffaoysa,3,5-bis 4-nitrophenoxy benzoic acid,3,5-bis 4-nitrophenoxy benzoic acid
16760376
C1=CC(=CC=C1[N+](=O)[O-])OC2=CC(=CC(=C2)C(=O)O)OC3=CC=C(C=C3)[N+](=O)[O-]
This item is not returnable. View return policy
173550-33-9
C19H12N2O8
396.311
JOSXKPZXMVHRKU-UHFFFAOYSA-N
3,5-bis 4-nitrophenoxy benzoic acid,gamma-secretase modulator, cw,cw,acmc-1cuip,josxkpzxmvhrku-uhfffaoysa,3,5-bis 4-nitrophenoxy benzoic acid,3,5-bis 4-nitrophenoxy benzoic acid
16760376
C1=CC(=CC=C1[N+](=O)[O-])OC2=CC(=CC(=C2)C(=O)O)OC3=CC=C(C=C3)[N+](=O)[O-]

γ-secretase inhibitor

Inhibitor of γ-secretase; causes a decrease in the released levels of Aβ42 and notch-1 Aβ-like peptide 25 (Nβ25).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
Formula Weight 396.31
Percent Purity >99%
Chemical Name or Material Compound W

Recommended Storage : Store at +4°C