Learn More
Chemglass Life Sciences PFA 90° Adapter, 1/2" Compression to 1/4" NPT
SDP
Supplier:  Chemglass Life Sciences CG1975E20
Encompass_Preferred
PFA 90° Adapter, 1/2" Compression to 1/4" NPT. PFA 90° adapter, compression to male NPT fittings are ideally suited for use in corrosive environments and chemical applications. The fitting design yields a leak-tight connection and requires no special tools to assemble. The connection is achieved by merely inserting the tubing into the fitting assembly and tightening the nut.
Catalog No. 50-143-615
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.