Learn More
Fisherbrand™ Computer Data Acquisition System for Digital Barometer Products for Science Education
Keep track of results for thermometers, hygrometers, barometers, light meters, anemometers, manometers, dissolved oxygen, conductivity and more. Fisher Scientific™ Computer Data Acquisition System for Digital Barometer is for use with Fisher Scientific™ Traceable™ Digital Barometer.
Catalog No. S2-509-9
Add to cart
Description
Description
Keep track of results for thermometers, hygrometers, barometers, light meters, anemometers, manometers, dissolved oxygen, conductivity and more. Fisher Scientific™ Computer Data Acquisition System for Digital Barometer is for use with Fisher Scientific™ Traceable™ Digital Barometer.
  • Allows up to eight unique Traceable™ meters to simultaneously capture data and send it to directly to email to any smartphone or computer
  • Permanently capture readings; keep track of results
  • Readings are saved to a file that can be viewed, printed as-is, or merged in a report or spreadsheet format
  • Data continues to be collected in background even when user works on other software programs
  • 24/7 audible/visual alarm reminds user to check on data
Specifications
Specifications
Computer Data Acquisition System
Education
CD, 6 ft. cable
Easily keeps track of results for thermometers, hygrometers, barometers, light meters, anemometers, manometers, dissolved oxygen, conductivity and more