Thermo Scientific™ Relative Humidity System Options for Environmental Chambers and Incubators
Address specific requirements to the relative humidity system of your chamber.
Manufacturer: Thermo Scientific™ 1900391
 View more versions of this product
Catalog No. 13-762-731
  Check Availability
Add to Cart
Disclaimers:
The heatless dryer requires an air supply of 6.2 bar (90 PSI) at 17 m3/h (10 CFM) compressor, at minimum. It is recommended that the air supply be greater than the minimum required by the dryer.
    Specifications
    For Use With (Equipment) Environmental Chambers (3948 and 3907)
    Description Condensate Evaporator; includes separate line cord (240V)
    Installation Customer Installed
    Use Thermo Scientific™ Relative Humidity System Options for Environmental Chambers to address specific requirements to the relative humidity system.
  • Description & Specifications
Related Products
Product Content Feedback