Learn More
Squishy Circuits™ Conductivity Kit Products for Science Education
Squishy Circuits™ Kit allows for the exploration of electrical circuits with conductive and insulative dough.
$15.00 - $275.00
Specifications
Min. Age 8
Quantity 1 Kit
View More Specs
Alert:
Choking Hazard
Products 5
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S29668 Products for Science Education
View Documents
Squishy Circuits™
DELUXE
Deluxe Kit
Each for $75.00
Add to cart
S29670 Products for Science Education
View Documents
Squishy Circuits™
DOUGH
Dough Kit
Each for $20.00
Add to cart
S29669 Products for Science Education
View Documents
Squishy Circuits™
GROUP
Group Kit
Each for $275.00
Add to cart
S29666 Products for Science Education
View Documents
Squishy Circuits™
LITE
Lite Kit
Each for $15.00
Add to cart
S29667 Products for Science Education
View Documents
Squishy Circuits™
STANDARD
Standard Kit
Each for $35.00
Add to cart
Description
Description
  • Deluxe Kit - The most complete kit that includes a deluxe battery holder, which has a knob that allows your lights to blink and buzzers to beep at different speeds
    • Contents: 1 x Deluxe Battery Holder, 1 x Piezoelectric Buzzer, 1 x Motor with Fan, 1 x Mechanical Buzzer, 1 x Switch, 40 x LEDs (5 each or Red, Green, White, Yellow, Blue, Purple, Orange, and Color Changing), 1 x Set of Dough Sculpting Tools, 1 x Rolling Pin, 1 x 7 oz. Container of White Insulating Dough, 7 x 7 oz. Containers of Conductive Dough (in Red, Blue, and Green, Yellow, Orange, Purple, and Black)
  • Dough Kit - Designed to make play dough science projects fun through an array of different colors
    • Contents: 7 containers of conductive dough and 1 container of white insulating dough
    • Each container holds 7 ounces
    • The dough is safe, non-toxic, and does contain gluten
    • The conductive dough dries out if left out of the container
    • The insulating dough does not dry out
  • Group Kit - Ideal for classroom groups of up to 30 peoples
    • Contents: 10 x Battery Holders (40 AA Batteries not Included), 10 x Piezoelectric Buzzers, 10 x Motors with Fan Blades, 10 x Switches, 80 x LEDs (10 each in Red, Green, White, Yellow, Orange, Purple, Blue, and Color Changing)
    • Kit does not come with dough
  • Lite Kit - Basic kit that is ideal for classrooms and events where every participant needs their own battery pack
    • Contents: 1 x Battery Holder, 15 x LEDs (5 each or Red, Green, Blue), 1 x 7 oz. Container of Conductive Dough (colors vary)
  • Standard Kit - Includes everything needed to get started with more advanced projects
    • Contents: 1 x Battery Holder, 1 x Piezoelectric Buzzer, 25 x LEDs (5 each or Red, Green, White, Yellow, Blue), 1 x 3.5 oz. Container of Insulating Dough, 3 x 3.5 oz. Containers of Conductive Dough (in Red, Green, and Blue)
Specifications
Specifications
8
1 Kit