Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conductivity Standard - 1,412 mhos/cm, MilliporeSigma™ Supelco™

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

$66.10 - $88.20

Specifications

Grade Certified Reference Material
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-6314
View Documents
MilliporeSigma Supelco
COND1412500ML
500 mL
Each for $66.10
Add to cart
 
11-100-4689
View Documents
MilliporeSigma Supelco
COND14121L
1 L
Each for $88.20
Add to cart
 
Description

Description

Featured Industries: Environmental
Specifications

Specifications

Certified Reference Material
Safety and Handling

Safety and Handling

Recommended Storage : Room Temp

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit