Learn More
Conductivity Standard, 2,765 umho/cm, Spectrum™
Manufacturer:  Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner C5821LT
Catalog No. 18-609-904
Add to cart
Specifications
Specifications
7447-40-7, 7732-18-5
Chemical Solution
1 L
Clear, Colorless
Poly Bottle
2765 ± 50 umho/cm
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.