Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conductivity Standard, 300,000 μS/cm at 25°C (300 mS/cm), Reagents

Catalog No. p-200021912
Click to view available options
Quantity:
500 mL

Conductivity Standard, 300,000 μS/cm at 25°C (300 mS/cm), Reagent, use for Conductivity meter calibration. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Conductivity Standard, 300,000 μS/cm at 25°C (300 mS/cm)
Packaging Poly Bottle
CAS 7732-18-5, 7647-01-0
Quantity 500 mL
Specific Gravity 1.014
Physical Form Liquid
Technique Calibration of Conductivity Instrument
Color Colorless
pH 0
For Use With (Equipment) Conductivity Meter
Solubility Completely miscible
Grade Reagent
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.