Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Specifications
Chemical Name or Material | Ricca Chemical Company Conductivity Standard |
---|---|
CAS | 7647-14-5, 7732-18-5 |
CAS Min % | 98.0, 0.01 |
CAS Max % | 100.0, 0.01 |
Specifications
Ricca Chemical Company Conductivity Standard | |
7647-14-5, 7732-18-5 | |
98.0, 0.01 | |
100.0, 0.01 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title