Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conductivity/TDS Standard, 10,000 μmho/cm (10 mS/cm), Reagents

Catalog No. 01335451
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
20 L
Catalog No. Quantity
01-335-451 4 L
01-335-454 100 mL
01-335-453 1 L
01-335-452 20 L
01-335-450 500 mL
5 options

Catalog No. 01-335-451

Supplier: Reagents Holdings Llc CC9700004A

Only null left
Add to Cart
Add to Cart

Conductivity/TDS Standard, 10,000 μmho/cm (10 mS/cm), NIST Traceable, use for Conductivity meter calibration. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.

Specifications

Chemical Name or Material Conductivity/TDS Standard, 10,000 μmho/cm (10 mS/cm)
Packaging Poly Bottle
CAS 7732-18-5, 7447-40-7
Specific Gravity 1.004
Physical Form Liquid
Molecular Formula KCl/H2O
Technique Calibration of Conductivity Instrument
Color Colorless
pH 5.4 to 8.6
Boiling Point 100°C
For Use With (Equipment) Conductivity Meter
Solubility Completely miscible
Quantity 4 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.