Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conductivity/TDS Standard, 1409 μS/cm, Reagents

Catalog No. 01335485
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
200 L
Catalog No. Quantity
01-335-485 20 L
01-335-488 10 L
01-335-487 1 L
01-335-486 200 L
01-335-484 4 L
5 options

Catalog No. 01-335-485

Supplier: Reagents Holdings Llc CC91390020F

Only null left
Add to Cart
Add to Cart

Conductivity/TDS Standard, 1409 μS/cm, NIST Traceable, use for Conductivity meter calibration. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.

Specifications

Chemical Name or Material Conductivity/TDS Standard, 1409 μS/cm
Packaging Cubitainer
CAS 7732-18-5, 7447-40-7
Specific Gravity 1.0004
Physical Form Liquid
Molecular Formula KCl/H2O
Technique Calibration of Conductivity Instrument
Color Colorless
pH 5.4 to 8.6
Boiling Point 100°C
For Use With (Equipment) Conductivity Meter
Solubility Completely miscible
Quantity 20 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.