Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Conductivity Standard, 200 μS/cm at 25°C (95 ppm TDS as NaCl), Ricca Chemical
Supplier: Ricca Chemical Company 223832
Specifications
Liquid | |
7647-14-5, 7732-18-5 | |
MFCD00003477 | |
[Na+].[Cl-] | |
58.44 | |
CHEBI:26710 | |
Colorless | |
Standard Solution | |
95ppm as NaCl |
Conductivity/TDS Standard | |
ClNa | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
sodium chloride | |
5234 | |
Natural Poly Bottle | |
1 L | |
Conductivity and TDS Standard |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction