Learn More
Brady™ Conduit & Voltage Labels - Self-Sticking Polyester
Self-Sticking Polyester (B-302) is a flexible polymer plastic over-laminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals.
Manufacturer:  Brady™ 86786
 View more versions of this product
Catalog No. 19-107-780
Add to cart
Description
Description
  • Weathers up to 8 years outdoors, even longer indoors, and withstands wide temperature variations (up to 230°F)
  • Flexible material conforms to most surfaces while protected graphics hold up to harsh environments, abrasion, and repeated spills and cleanings
  • Permanent, pressure sensitive adhesive forms a long-lasting bond to even the most difficult surfaces and shapes
  • Cold temperature adhesive allows the signage to be applied at 0°F (-18°C)
Specifications
Specifications
Self-Adhesive
Not Autoclavable
Black/Red on White
0.006 in.
Label
Gloss
Pack of 5 Labels
1
3.5H x 5W x 0.006 in. D
92.71 mm
Polyester
Pack of 5 Labels
Safety
Permanent Acrylic
ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145
220 VOLTS
3.5 in. H x 5 in. W x 0.006 in. D
3.5 in.
Pack of 5 Labels
Rectangle
5 in.
English
Label