Learn More
Brady™ Conduit & Voltage Labels - Self-Sticking Polyester
Self-Sticking Polyester (B-302) is a flexible polymer plastic over-laminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals.
$15.75
Products 6
Catalog Number Mfr. No. Display Text Price Quantity & Availability  
Catalog Number Mfr. No. Display Text Price Quantity & Availability  
19-107-775 View Documents Brady™
86221
_____ VOLTS, OSHA Pack of 5 for $15.75
Add to cart
19-107-777 View Documents Brady™
86783
480 VOLTS Pack of 5 for $15.75
Add to cart
19-107-779 View Documents Brady™
86785
240 VOLTS Pack of 5 for $15.75
Add to cart
19-107-778 View Documents Brady™
86784
120 VOLTS Pack of 5 for $15.75
Add to cart
19-107-781 View Documents Brady™
86870
_____ VOLTS, ANSI Pack of 5 for $15.75
Add to cart
19-107-780 View Documents Brady™
86786
220 VOLTS Pack of 5 for $15.75
Add to cart
Description
Description
  • Weathers up to 8 years outdoors, even longer indoors, and withstands wide temperature variations (up to 230°F)
  • Flexible material conforms to most surfaces while protected graphics hold up to harsh environments, abrasion, and repeated spills and cleanings
  • Permanent, pressure sensitive adhesive forms a long-lasting bond to even the most difficult surfaces and shapes
  • Cold temperature adhesive allows the signage to be applied at 0°F (-18°C)