Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Bradycone™ Warning System - 35 in H

Use our Bradycone™ Warning System for any of your hazardous warning needs

Manufacturer:  Brady™ 103680

 View more versions of this product

Catalog No. 19-104-399


Add to cart

Description

Description

  • Cones are four-sided, so your message can't be missed
Specifications

Specifications

Each
Yellow
35 in.
CAUTION/PRECAUCION WET FLOOR/PISO MOJADO
Cone
Cone
Plastic
323.85 mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit