Alfa Aesar™ Congo Red, Dye content»80%

Manufacturer: Alfa Aesar™ 4456606

Quantity 5g

 View more versions of this product

Catalog No. AA4456606

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 573-58-0
    Molecular Formula C32H22N6Na2O6S2
    Formula Weight 696.67
    MDL Number MFCD00004028
    Beilstein 3894858
    Merck Index 14,2498
    Synonym C.I. 22120
    Chemical Name or Material Congo Red, Dye content ≈80%

    Specifications

    Formula Weight 696.67
    Health Hazard 3 GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Merck Index 14,2498
    Synonym C.I. 22120
    Odor Odorless
    Physical Form Powder
    MDL Number MFCD00004028
    Beilstein 3894858
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H350-H361
    May cause cancer.
    Suspected of damaging fertility or the unborn child.
    RTECS Number QK1400000
    Quantity 5g
    TSCA TSCA
    Molecular Formula C32H22N6Na2O6S2
    Melting Point >360°C
    CAS 573-58-0
    Chemical Name or Material Congo Red, Dye content ≈80%

    GHS H Statement
    H350-H361
    May cause cancer.
    Suspected of damaging fertility or the unborn child.

    GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit