Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Congo Red Indicator, 0.1% Aqueous, Reagents
Click to view available options
Quantity:
100 mL
500 mL
1 L
Specifications
Specifications
Chemical Name or Material | Congo Red Indicator, 0.1% Aqueous |
Boiling Point | 100°C |
CAS | 7732-18-5, 573-58-0 |
Synonym | Direct Red 28 |
Percent Purity | 0.1% |
Grade | Reagent |
Packaging | Glass Bottle |
Specific Gravity | 1.0008 |
Color | Red |
Quantity | 1 L |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction