Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Congo Red, Spectrum™

Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Amber Glass Bottle
573-58-0
573-58-0
CAS Max % 1
Grade Reagent
Packaging Amber Glass Bottle
Quantity 500 g

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.