Learn More
Congo Red TS, (U.S.P. Test Solution), Spectrum™
Click to view available options
Packaging:
Amber Glass Bottle
C-292, 573-58-0
Spectrum™ Congo Red TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
Packaging Amber Glass Bottle