Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Congo Red TS, (U.S.P. Test Solution), Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Amber Glass Bottle
573-58-0,7732-18-5
C32H22N6Na2O6S2
696.66
IQFVPQOLBLOTPF-UHFFFAOYSA-L
disodium 4-amino-3-(2-{4'-[2-(1-amino-4-sulfonatonaphthalen-2-yl)diazen-1-yl]-[1,1'-biphenyl]-4-yl}diazen-1-yl)naphthalene-1-sulfonate
[Na+].[Na+].NC1=C2C=CC=CC2=C(C=C1N=NC1=CC=C(C=C1)C1=CC=C(C=C1)N=NC1=CC(=C2C=CC=CC2=C1N)S([O-])(=O)=O)S([O-])(=O)=O
573-58-0,7732-18-5
C32H22N6Na2O6S2
696.66
IQFVPQOLBLOTPF-UHFFFAOYSA-L
disodium 4-amino-3-(2-{4'-[2-(1-amino-4-sulfonatonaphthalen-2-yl)diazen-1-yl]-[1,1'-biphenyl]-4-yl}diazen-1-yl)naphthalene-1-sulfonate
[Na+].[Na+].NC1=C2C=CC=CC2=C(C=C1N=NC1=CC=C(C=C1)C1=CC=C(C=C1)N=NC1=CC(=C2C=CC=CC2=C1N)S([O-])(=O)=O)S([O-])(=O)=O

C-292, 573-58-0

Spectrum™ Congo Red TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
CAS Max % 0.1%,99.9%
Packaging Amber Glass Bottle
Quantity 500 mL