Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Conical Tubes
GREENER_CHOICE

Ideally suited for cell biology applications and sample preparation protocols in microbiology and molecular biology laboratories.

Supplier:  Eppendorf™ 0030122178

 View more versions of this product

Catalog No. 13-690-085


Add to cart

Description

Description

  • Premium USP class 6 raw material
  • No use of slip agents, plasticizers, and biocides during manufacturing
  • Ensures high sample integrity, g-Safe™ centrifugation stability, optimal sample and pellet visibility
  • Cap with flattened and grooved sides supports user-friendly ergonomic handling
  • Enables stable upright positioning, and minimizes contamination risk
  • Reliable and safe labelling due to large writing area on the tube and due to flat and light-colored cap
  • Precise dimensions enable maximum compatibility with centrifuge rotors, mixers and shakers

Applications

  • Convenient and safe cell culture applications
  • Sample preparation in microbiology and molecular biology labs
  • Storage of samples and solution with larger volumes
  • Preparation of mastermixes and buffers
Specifications

Specifications

HDPE
Screw Cap, flattened and grooved sides
19,500 x g
Writing area
Bag
-86°C to -100°C
50 mL
Pyrogen, DNase, RNase, human and bacterial DNA free
Light Gray
Colorless
Polypropylene
20 × 25
Cs. of 500
Sterile
30 mm
Conical
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit