Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eppendorf™ Conical Tubes
Ideally suited for cell biology applications and sample preparation protocols in microbiology and molecular biology laboratories.
Supplier: Eppendorf™ 0030122259
Description
- Premium USP class 6 raw material
- No use of slip agents, plasticizers, and biocides during manufacturing
- Ensures high sample integrity, g-Safe™ centrifugation stability, optimal sample and pellet visibility
- Cap with flattened and grooved sides supports user-friendly ergonomic handling
- Enables stable upright positioning, and minimizes contamination risk
- Reliable and safe labelling due to large writing area on tube and due to flat and light-colored cap
- Precise dimensions enable maximum compatibility with centrifuge rotors, mixers and shakers
Applications
- Convenient and safe cell culture applications
- Sample preparation in microbiology and molecular biology labs
- Storage of samples and solution with larger volumes
- Preparation of master mixes and buffers
Specifications
HDPE | |
Screw Cap, flattened and grooved sides | |
19,500 x g | |
Writing area | |
Individually Wrapped | |
Non-sterile | |
17 mm | |
Conical |
Light Colored | |
Colorless | |
Polypropylene | |
100 | |
-86°C to -100°C | |
15 mL | |
Forensic DNA Grade |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title