Learn More
MilliporeSigma™ Supelco™ Conikal™ Nebulizer, 2 mL/min. with EzyFit EZT-075 & EzyLok, Gas Flow: 1 mL/min.
Includes EZT-075 EzyFit Connector with 0.75 mm ID x 1.3 mm OD x 700 mm L tubing (1 unit) and EL-1 EzyLok Connector Kit
Manufacturer:  MilliporeSigma™ Supelco™ GXAR301FC2E1EA
Catalog No. 11-100-7600
Add to cart
Description
Description
  • This nebulizer supersedes AR30-1-FC2
  • Suggested Gas Flow 1 L/min.
  • Suggested Sample Uptake 2 mL/min.
Specifications
Specifications
Agilent/Varian Liberty Series II Axial, Analytik Jena/Bruker, Varian Aurora M90, 800-MS Series, ICP-MS, Leeman/Teledyne Leeman Series Axial/DV, Prodigy, Profile, Leeman/Teledyne Leeman Series Radial, Prodigy, Profile, PerkinElmer Elan 500, 5000, 6000, 9000, PerkinElmer Elan 6100 DRC, DRC II, DRC-e, PerkinElmer Optima 3000 RL, 3100 RL, 3200 RL and SII NanoTechlology/Seiko SPS/SPQ Series
Spectrometry
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Conikal is a trademark of Glass Expansion Pty Ltd